Please login to your online account to display your discounted pricing

γ-Phenyl-γ-butyrolactone, 97%, ACROS Organics™

$102.03 - $102.03

Chemical Identifiers

Chemical Name or Material γ-Phenyl-γ-butyrolactone
Name Note 97%
CAS 1008-76-0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C10H10O2
Formula Weight 162.19
MDL Number MFCD00005398
Synonym 4, 5-Dihydro-5-phenyl-2(3H)-furanone
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC175210250

  SDS
thermo fisher scientific
175210250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC175210050

  SDS
thermo fisher scientific
175210050
Glass bottle 5g Each for $102.03
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid After Melting
Formula Weight 162.19
Quantity 25g
Color Clear colorless to yellow
EINECS Number 213-761-0
TSCA TSCA
RTECS Number LU3690000
Density 1.1500g/mL
Refractive Index 1.5415 to 1.5435 (20°C, 589nm)
Name Note 97%
Molecular Formula C10H10O2
CAS 1008-76-0
Synonym 4, 5-Dihydro-5-phenyl-2(3H)-furanone
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material γ-Phenyl-γ-butyrolactone
Assay Percent Range 97%
MDL Number MFCD00005398