missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

γ-Phenyl-γ-butyrolactone, 97%, ACROS Organics™

Manufacturer:  Acros Organics 175210050

 View more versions of this product

Catalog No. AC175210050



Specifications

Specifications

γ-Phenyl-γ-butyrolactone
>112°C
Glass bottle
306.0°C
36.0°C to 37.0°C
97%
97%
MFCD00005398
AEUULUMEYIPECD-UHFFFAOYSA-N
5-phenyloxolan-2-one
13884
Liquid After Melting
1.1500g/mL
Authentic
1.5415 to 1.5435
Colorless to Yellow
5g
1008-76-0
C10H10O2
gamma-phenyl-gamma-butyrolactone, 4-phenylbutanolide, 4-phenyl-4-butanolide, 4-phenylbutyrolactone, 2 3h-furanone, dihydro-5-phenyl, 5-phenyldihydrofuran-2 3h-one, 4-phenyl-4-hydroxybutanoic acid lactone, 4,5-dihydro-5-phenyl-2 3h-furanone, .gamma.-phenyl-.gamma.-butyrolactone, .gamma.-phenylbutyrolactone
C1CC(=O)OC1C2=CC=CC=C2
162.188
162.19
97%

Chemical Identifiers

1008-76-0
162.188
AEUULUMEYIPECD-UHFFFAOYSA-N
13884
C1CC(=O)OC1C2=CC=CC=C2
C10H10O2
MFCD00005398
gamma-phenyl-gamma-butyrolactone, 4-phenylbutanolide, 4-phenyl-4-butanolide, 4-phenylbutyrolactone, 2 3h-furanone, dihydro-5-phenyl, 5-phenyldihydrofuran-2 3h-one, 4-phenyl-4-hydroxybutanoic acid lactone, 4,5-dihydro-5-phenyl-2 3h-furanone, .gamma.-phenyl-.gamma.-butyrolactone, .gamma.-phenylbutyrolactone
5-phenyloxolan-2-one
Safety and Handling

Safety and Handling

EINECSNumber : 213-761-0

RTECSNumber : LU3690000

TSCA : TSCA

SDS Documents