Learn More
Phenyl bromoacetate, 98%, Thermo Scientific Chemicals
$58.74 - $410.67
Chemical Identifiers
CAS | 620-72-4 |
---|---|
Molecular Formula | C8H7BrO2 |
Molecular Weight (g/mol) | 215.046 |
MDL Number | MFCD00192391 |
InChI Key | UEWYUCGVQMZMGY-UHFFFAOYSA-N |
Synonym | phenyl bromoacetate, acetic acid, bromo-, phenyl ester, bromoacetic acid phenyl ester, bromoacetic acid, phenyl ester, phenylbromessigester, phenyl bromoacetate #, acmc-209mzb, phenyl 2-bromanylethanoate, ksc352o8b |
PubChem CID | 564919 |
IUPAC Name | phenyl 2-bromoacetate |
SMILES | C1=CC=C(C=C1)OC(=O)CBr |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6046406
|
Thermo Scientific Chemicals
H6046406 |
5 g |
Each for $58.74
|
|
|||||
AAH6046418
|
Thermo Scientific Chemicals
H6046418 |
50 g |
Each for $410.67
|
|
|||||
Description
Phenyl bromoacetate may be employed as alkylation reagent in the preparation of 2-(phenoxycarbonyl)methyl triazoles. It may be used in the synthesis of the A-ring of cylindrospermopsin and in the synthesis of 2-{2-[1-(4-hydroxy-6-methyl-2-oxo-2H-pyran-3-yl)ethylidene]hydrazinyl}-1,3-thiazolidin-4-one3, 2-{2-[1-(4-hydroxy-6-methyl-2-oxo-2H-pyran-3-yl)ethylidene]hydrazinyl}-3-methyl-1,3-thiazolidin-4-one3, 3-ethyl-2-{2-[1-(4-hydroxy-6-methyl-2-oxo-2H-pyran-3-yl)ethylidene]hydrazinyl}-1,3-thiazolidin-4-one3 and 2-{2-[1-(4-hydroxy-6-methyl-2-oxo-2H-pyran-3-yl)ethylidene] hydrazinyl}-3-phenyl-1,3-thiazolidin-4-one3.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPhenyl bromoacetate may be employed as alkylation reagent in the preparation of 2-(phenoxycarbonyl)methyl triazoles. It may be used in the synthesis of the A-ring of cylindrospermopsin and in the synthesis of 2-{2-[1-(4-hydroxy-6-methyl-2-oxo-2H-pyran-3-yl)ethylidene]hydrazinyl}-1,3-thiazolidin-4-one3, 2-{2-[1-(4-hydroxy-6-methyl-2-oxo-2H-pyran-3-yl)ethylidene]hydrazinyl}-3-methyl-1,3-thiazolidin-4-one3, 3-ethyl-2-{2-[1-(4-hydroxy-6-methyl-2-oxo-2H-pyran-3-yl)ethylidene]hydrazinyl}-1,3-thiazolidin-4-one3 and 2-{2-[1-(4-hydroxy-6-methyl-2-oxo-2H-pyran-3-yl)ethylidene] hydrazinyl}-3-phenyl-1,3-thiazolidin-4-one3.
Solubility
Soluble in ethanol and ether, insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under inert gas.
Chemical Identifiers
620-72-4 | |
215.046 | |
UEWYUCGVQMZMGY-UHFFFAOYSA-N | |
564919 | |
C1=CC=C(C=C1)OC(=O)CBr |
C8H7BrO2 | |
MFCD00192391 | |
phenyl bromoacetate, acetic acid, bromo-, phenyl ester, bromoacetic acid phenyl ester, bromoacetic acid, phenyl ester, phenylbromessigester, phenyl bromoacetate #, acmc-209mzb, phenyl 2-bromanylethanoate, ksc352o8b | |
phenyl 2-bromoacetate |
Specifications
620-72-4 | |
1.508 | |
113°C (235°F) | |
BrCH2CO2C6H5 | |
5 g | |
Soluble in ethanol and ether,insoluble in water. | |
C1=CC=C(C=C1)OC(=O)CBr | |
215.046 | |
215.04 | |
Phenyl bromoacetate |
31°C to 33°C | |
134°C | |
C8H7BrO2 | |
MFCD00192391 | |
phenyl bromoacetate, acetic acid, bromo-, phenyl ester, bromoacetic acid phenyl ester, bromoacetic acid, phenyl ester, phenylbromessigester, phenyl bromoacetate #, acmc-209mzb, phenyl 2-bromanylethanoate, ksc352o8b | |
UEWYUCGVQMZMGY-UHFFFAOYSA-N | |
phenyl 2-bromoacetate | |
564919 | |
98% |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only