missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenyl benzoate, 99%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 162061000

 View more versions of this product

Catalog No. AC162061000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

1mg KOH/g max.
Authentic
298.0°C to 299.0°C
68.0°C to 70.0°C
99%
98.5% min. (GC)
C6H5CO2C6H5
09, 116
benzoic acid phenyl, benzoic acid phenyl ester, benzoic acid, phenyl ester, diphenylcarboxylate, dsstox_cid_28185, dsstox_gsid_48210, dsstox_rid_82744, phenol, benzoate, phenylbenzoate, unii-b8a3wvz590
FCJSHPDYVMKCHI-UHFFFAOYSA-N
phenyl benzoate
7169
198.22
99%
Phenyl benzoate
Plastic bottle
White
100g
93-99-2
C13H10O2
MFCD00003072
15, 7388
Solubility in water: insoluble. Other solubilities: soluble in ethanol,ethyl ether and chloroform
C1=CC=C(C=C1)C(=O)OC2=CC=CC=C2
198.22
CHEBI:86919
Fine Crystalline Powder

Chemical Identifiers

93-99-2
198.22
FCJSHPDYVMKCHI-UHFFFAOYSA-N
7169
phenyl benzoate
C13H10O2
MFCD00003072
benzoic acid phenyl, benzoic acid phenyl ester, benzoic acid, phenyl ester, diphenylcarboxylate, dsstox_cid_28185, dsstox_gsid_48210, dsstox_rid_82744, phenol, benzoate, phenylbenzoate, unii-b8a3wvz590
CHEBI:86919
C1=CC=C(C=C1)C(=O)OC2=CC=CC=C2
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Con

GHS Signal Word: Warning

EINECSNumber : 202-293-2

RTECSNumber : DH6299500

TSCA : TSCA

SDS Documents