Please login to your online account to display your discounted pricing

Alfa Aesar™ Phenyl benzoate, 99%

$25.85 - $190.86

Chemical Identifiers

Chemical Name or Material Phenyl benzoate
CAS 93-99-2
Molecular Formula C13H10O2
Formula Weight 198.22
MDL Number MFCD00003072
Merck Index 14,7275
Synonym Benzoic acid phenyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2298614

  SDS
alfa aesar™
B2298614
25g Each for $25.85

AAB2298622

  SDS
alfa aesar™
B2298622
100g Each for $57.73

AAB2298636

  SDS
alfa aesar™
B2298636
500g Each for $190.86
Description & Specifications

Specifications

Formula Weight 198.22
Chemical Name or Material Phenyl benzoate
TSCA TSCA
Assay 99%
Molecular Formula C13H10O2
MDL Number MFCD00003072
CAS 93-99-2
Merck Index 14,7275
Synonym Benzoic acid phenyl ester
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number DH6299500
Boiling Point 298° to 299°C
Melting Point 68° to 70°C
Quantity 25g

Applications
Phenyl benzoate was used in the synthesis of soluble polyimides using dianhydride/diamine derivatives. It undergoes Fries rearrangement catalyzed by heteropoly acids to yield the acylated phenols and esters. Phenyl benzoate is considered an excellent starting material for the production of optical components, particularly high quality lenses for still and motion picture cameras.

Solubility
Soluble in ethanol, ethyl ether and chloroform. Insoluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.