missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Phenyl acrylate, 97%

Manufacturer:  Alfa Aesar™ L1181914

 View more versions of this product

Catalog No. AAL1181914



Description

Description

Applications
Phenyl acrylate reacts with benzaldehyde to produce 2-(hydroxy-phenyl-methyl)-acrylic acid phenyl ester.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents. Store at 4°C. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

Phenyl acrylate
25g
C9H8O2
UN3082
phenyl acrylate, phenylacrylate, 2-propenoic acid, phenyl ester, phenol acrylate, phenyl-acrylate, acrylic acid phenyl ester, phenyl acrylate 5g, 2-propenoic acid,phenyl ester, acrylic acid phenyl ester, stabilized with bht
C=CC(=O)OC1=CC=CC=C1
148.161
148.16
89°C to 90°C (12mmHg)
937-41-7
MFCD00048145
2041124
WRAQQYDMVSCOTE-UHFFFAOYSA-N
phenyl prop-2-enoate
61242
97%

Chemical Identifiers

937-41-7
148.161
WRAQQYDMVSCOTE-UHFFFAOYSA-N
61242
C=CC(=O)OC1=CC=CC=C1
C9H8O2
MFCD00048145
phenyl acrylate, phenylacrylate, 2-propenoic acid, phenyl ester, phenol acrylate, phenyl-acrylate, acrylic acid phenyl ester, phenyl acrylate 5g, 2-propenoic acid,phenyl ester, acrylic acid phenyl ester, stabilized with bht
phenyl prop-2-enoate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.

TSCA : TSCA

Documents