missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Phenyl acetoacetate on polystyrene, ca 3 mmol/g

$320.12 - $1,208.70

Chemical Identifiers

MDL Number MFCD03792524
Synonym (4-Acetylacetoxy)polystyrene; Activated ketone, polymer-bound
Chemical Name or Material Phenyl acetoacetate on polystyrene
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1959903 SDS Alfa Aesar™
L1959903
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAL1959906 SDS Alfa Aesar™
L1959906
5g Each for $320.12
AAL1959914 SDS Alfa Aesar™
L1959914
25g Each for $1,208.70
Specifications Description & Specifications

Chemical Identifiers

MFCD03792524
(4-Acetylacetoxy)polystyrene; Activated ketone, polymer-bound
Phenyl acetoacetate on polystyrene

Specifications

MFCD03792524
1g
Phenyl acetoacetate on polystyrene
(4-Acetylacetoxy)polystyrene; Activated ketone, polymer-bound
≈3mmol/g