missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Phenyl acetate, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 148770010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Phenyl acetate | |
122-79-2 | |
196.0°C | |
Authentic | |
Glass bottle | |
1.5010 to 1.5030 | |
MFCD00008699 | |
06,152 | |
1.07 | |
acetic acid phenyl ester, phenol acetate, acetylphenol, acetic acid, phenyl ester, acetyl phenol, acetoxybenzene, acetates, acetic acid,phenyl ester, fenylester kyseliny octove, fenylester kyseliny octove czech | |
IPBVNPXQWQGGJP-UHFFFAOYSA-N | |
phenyl acetate | |
31229 | |
136.15 |
97% | |
1.0700g/mL | |
76°C | |
96% min. (GC) | |
C8H8O2 | |
CH3CO2C6H5 | |
1kg | |
01,829 | |
15,7379 | |
Solubility in water: ca 4g/L (25°C). Other solubilities: miscible with alcohol,chloroform,ether,soluble in glacial acetic acid | |
CC(=O)OC1=CC=CC=C1 | |
136.15 | |
CHEBI:8082 | |
97% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning
EINECSNumber : 204-575-0
RTECSNumber : AJ2800000
TSCA : TSCA
RUO â Research Use Only