Please login to your online account to display your discounted pricing

Alfa Aesar™ Phenyl 4-[(R)-alpha-(4-chlorophenyl)benzyl]piperazine-1-carboxylate, 98%

Chemical Identifiers

Chemical Name or Material Phenyl 4-[(R)-alpha-(4-chlorophenyl)benzyl]piperazine-1-carboxylate
CAS 1136010-90-6
Formula Weight 406.9
Synonym Phenyl 4-[(R)-(4-chlorophenyl)(phenyl)methyl]piperazine-1-carboxylate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6178403

 
alfa aesar™
H6178403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH61784MD

 
alfa aesar™
H61784MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 406.9
Chemical Name or Material Phenyl 4-[(R)-alpha-(4-chlorophenyl)benzyl]piperazine-1-carboxylate
Physical Form Solid
Assay 98%
CAS 1136010-90-6
Synonym Phenyl 4-[(R)-(4-chlorophenyl)(phenyl)methyl]piperazine-1-carboxylate
Quantity 1g