Please login to your online account to display your discounted pricing

Phenyl(4-piperidyl)methanone hydrochloride, 97%

Chemical Identifiers

Chemical Name or Material Phenyl(4-piperidyl)methanone hydrochloride
Assay 97%
Assay Percent Range 97%
Molecular Formula C12H15NO·ClH
Formula Weight 225.72
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SEW04290DA

  SDS
fisher scientific company
SEW04290DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 225.72
Quantity 1g
Assay 97%
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C12H15NO·ClH
CAS 25519-80-6
Chemical Name or Material Phenyl(4-piperidyl)methanone hydrochloride