Please login to your online account to display your discounted pricing

Phenyl(4-piperidyl)methanone hydrochloride, 97%


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  SEW04290DA

 View more versions of this product

Catalog No. SEW04290DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 225.72
Quantity 1g
Assay 97%
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C12H15NO·ClH
CAS 25519-80-6
Chemical Name or Material Phenyl(4-piperidyl)methanone hydrochloride

Specifications

Chemical Name or Material Phenyl(4-piperidyl)methanone hydrochloride
Assay 97%
Assay Percent Range 97%
Molecular Formula C12H15NO·ClH
Formula Weight 225.72