missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenyl(2,4,6-trimethoxyphenyl)iodonium p-Toluenesulfonate, TCI America™

Manufacturer:  TCI America P24121G

 View more versions of this product

Catalog No. P24121G



Specifications

Specifications

Phenyl(2,4,6-trimethoxyphenyl)iodonium p-Toluenesulfonate
179°C
936326-60-2
Phenyl(2,4,6-trimethoxyphenyl)iodonium Tosylate
CC1=CC=C(C=C1)S(=O)(=O)[O-].COC1=CC(=C(C(=C1)OC)[I+]C2=CC=CC=C2)OC
542.384
542.38
≥93.0% (HPLC)
White-Yellow
1g
C22H23IO6S
GGEMRLZODUITOC-UHFFFAOYSA-M
4-methylbenzenesulfonate;phenyl-(2,4,6-trimethoxyphenyl)iodanium
131697920
Crystal-Powder at 20°C

Chemical Identifiers

936326-60-2
542.384
Phenyl(2,4,6-trimethoxyphenyl)iodonium Tosylate
4-methylbenzenesulfonate;phenyl-(2,4,6-trimethoxyphenyl)iodanium
C22H23IO6S
GGEMRLZODUITOC-UHFFFAOYSA-M
131697920
CC1=CC=C(C=C1)S(=O)(=O)[O-].COC1=CC(=C(C(=C1)OC)[I+]C2=CC=CC=C2)OC
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

Documents