missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenyl 2,3,4,6-Tetra-O-acetyl-1-thio-beta-D-galactopyranoside, TCI America™

Manufacturer:  TCI America P147725G

 View more versions of this product

Catalog No. P147725G



Specifications

Specifications

Phenyl 2,3,4,6-Tetra-O-acetyl-1-thio-beta-D-galactopyranoside
70°C
24404-53-3
phenyl 2,3,4,6-tetra-o-acetyl-1-thio-beta-d-galactopyranoside, phenyl 2,3,4,6-tetra-o-acetyl-b-d-thiogalactopyranoside, 2r,3s,4s,5r,6s-2-acetoxymethyl-6-phenylthio tetrahydro-2h-pyran-3,4,5-triyl triacetate, 2r,3s,4s,5r,6s-3,4,5-tris acetyloxy-6-phenylsulfanyl oxan-2-yl methyl acetate, pubchem9897, phenyl2,3,4,6-tetra-o-acetyl-1-thio-beta-d-galactopyranoside, phenyl-tetra-o-acetyl-thio-beta-d-galactopyranoside, 1-phenylthio-1-deoxy-beta-d-galactopyranose tetraacetate, 1-deoxy-1-phenylthio-2,3,4,6-tetra-o-acetyl-beta-d-galactopyranose, phenyl 2,3,4,6-tetra-o-acetyl-1-thio-beta-d-galactopyranoside hplc
CC(=O)OCC1C(C(C(C(O1)SC2=CC=CC=C2)OC(=O)C)OC(=O)C)OC(=O)C
440.463
440.46
≥98.0% (HPLC)
White
25g
C20H24O9S
JCKOUAWEMPKIAT-CXQPBAHBSA-N
[(2R,3S,4S,5R,6S)-3,4,5-triacetyloxy-6-phenylsulfanyloxan-2-yl]methyl acetate
10950254
Crystal-Powder at 20°C

Chemical Identifiers

24404-53-3
440.463
phenyl 2,3,4,6-tetra-o-acetyl-1-thio-beta-d-galactopyranoside, phenyl 2,3,4,6-tetra-o-acetyl-b-d-thiogalactopyranoside, 2r,3s,4s,5r,6s-2-acetoxymethyl-6-phenylthio tetrahydro-2h-pyran-3,4,5-triyl triacetate, 2r,3s,4s,5r,6s-3,4,5-tris acetyloxy-6-phenylsulfanyl oxan-2-yl methyl acetate, pubchem9897, phenyl2,3,4,6-tetra-o-acetyl-1-thio-beta-d-galactopyranoside, phenyl-tetra-o-acetyl-thio-beta-d-galactopyranoside, 1-phenylthio-1-deoxy-beta-d-galactopyranose tetraacetate, 1-deoxy-1-phenylthio-2,3,4,6-tetra-o-acetyl-beta-d-galactopyranose, phenyl 2,3,4,6-tetra-o-acetyl-1-thio-beta-d-galactopyranoside hplc
[(2R,3S,4S,5R,6S)-3,4,5-triacetyloxy-6-phenylsulfanyloxan-2-yl]methyl acetate
C20H24O9S
JCKOUAWEMPKIAT-CXQPBAHBSA-N
10950254
CC(=O)OCC1C(C(C(C(O1)SC2=CC=CC=C2)OC(=O)C)OC(=O)C)OC(=O)C
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer

Documents