missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenyl 1-Hydroxy-2-naphthoate, TCI America™

$90.14 - $652.38

Chemical Identifiers

CAS 132-54-7
Molecular Formula C17H12O3
Molecular Weight (g/mol) 264.28
MDL Number MFCD00014310
InChI Key QHDYIMWKSCJTIM-UHFFFAOYSA-N
Synonym phenyl 1-hydroxy-2-naphthoate, 1-hydroxy-2-naphthoic acid phenyl ester, 2-naphthalenecarboxylic acid, 1-hydroxy-, phenyl ester, phenyl 1-hydroxy-2-naphthalenecarboxylate, unii-0749hk9b6b, phenyl-1-hydroxy-2-naphthate, phenyl-1-hydroxy-2-naphthoate, 1-hydroxy-2-naphthalenecarboxylic acid phenyl ester, aon phenyl ester, 1-hydroxy-2-naphthoic
PubChem CID 67236
IUPAC Name phenyl 1-hydroxynaphthalene-2-carboxylate
SMILES C1=CC=C(C=C1)OC(=O)C2=C(C3=CC=CC=C3C=C2)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H0797500G View Documents TCI America
H0797500G
500g Each for $652.38
 
H079725G View Documents TCI America
H079725G
25g Each for $90.14
 
Specifications

Chemical Identifiers

132-54-7
264.28
QHDYIMWKSCJTIM-UHFFFAOYSA-N
67236
C1=CC=C(C=C1)OC(=O)C2=C(C3=CC=CC=C3C=C2)O
C17H12O3
MFCD00014310
phenyl 1-hydroxy-2-naphthoate, 1-hydroxy-2-naphthoic acid phenyl ester, 2-naphthalenecarboxylic acid, 1-hydroxy-, phenyl ester, phenyl 1-hydroxy-2-naphthalenecarboxylate, unii-0749hk9b6b, phenyl-1-hydroxy-2-naphthate, phenyl-1-hydroxy-2-naphthoate, 1-hydroxy-2-naphthalenecarboxylic acid phenyl ester, aon phenyl ester, 1-hydroxy-2-naphthoic
phenyl 1-hydroxynaphthalene-2-carboxylate

Specifications

White-Yellow
500g
C17H12O3
phenyl 1-hydroxy-2-naphthoate, 1-hydroxy-2-naphthoic acid phenyl ester, 2-naphthalenecarboxylic acid, 1-hydroxy-, phenyl ester, phenyl 1-hydroxy-2-naphthalenecarboxylate, unii-0749hk9b6b, phenyl-1-hydroxy-2-naphthate, phenyl-1-hydroxy-2-naphthoate, 1-hydroxy-2-naphthalenecarboxylic acid phenyl ester, aon phenyl ester, 1-hydroxy-2-naphthoic
C1=CC=C(C=C1)OC(=O)C2=C(C3=CC=CC=C3C=C2)O
264.28
264.28
≥98.0% (GC,T)
95°C
132-54-7
MFCD00014310
QHDYIMWKSCJTIM-UHFFFAOYSA-N
phenyl 1-hydroxynaphthalene-2-carboxylate
67236
Crystal-Powder at 20°C
Phenyl 1-Hydroxy-2-naphthoate
Safety and Handling

Safety and Handling

TSCA : Yes