missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phentolamine Mesylate, TCI America™

Manufacturer:  TCI America P19851G

 View more versions of this product

Catalog No. P19851G



Specifications

Specifications

Phentolamine Mesylate
180°C
65-28-1
C18H23N3O4S
2-[N-(m-Hydroxyphenyl)-p-toluidinomethyl]imidazoline Methanesulfonate, Phentolamine Methanesulfonate
CC1=CC=C(C=C1)N(CC2=NCCN2)C3=CC(=CC=C3)O.CS(=O)(=O)O
377.459
377.46
White-Yellow
1g
≥98.0% (HPLC,T)
MFCD00134201
OGIYDFVHFQEFKQ-UHFFFAOYSA-N
3-[N-(4,5-dihydro-1H-imidazol-2-ylmethyl)-4-methylanilino]phenol;methanesulfonic acid
91430
Crystal-Powder at 20°C

Chemical Identifiers

65-28-1
377.459
OGIYDFVHFQEFKQ-UHFFFAOYSA-N
91430
CC1=CC=C(C=C1)N(CC2=NCCN2)C3=CC(=CC=C3)O.CS(=O)(=O)O
C18H23N3O4S
MFCD00134201
2-[N-(m-Hydroxyphenyl)-p-toluidinomethyl]imidazoline Methanesulfonate, Phentolamine Methanesulfonate
3-[N-(4,5-dihydro-1H-imidazol-2-ylmethyl)-4-methylanilino]phenol;methanesulfonic acid
Safety and Handling

Safety and Handling

RTECSNumber : SL5430000

TSCA : No

Documents