Please login to your online account to display your discounted pricing

Phenoxytrimethylsilane, +97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Phenoxytrimethylsilane
Assay 97.0% min.
Assay Percent Range 97+%
Molecular Formula C9H14OSi
Linear Formula C6H5OSi(CH3)3
Formula Weight 166.29g/mol
MDL Number MFCD00053666
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC388600050

  SDS
thermo fisher scientific
388600050
Glass bottle 5mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4760 to 1.4780 (20°C, 589nm)
Density 0.9256g/mL
Quantity 5mL
Color Clear colorless
Chemical Name or Material Phenoxytrimethylsilane
CAS 1529-17-5
Physical Form Clear colorless liquid
Assay 97.0% min.
Assay Percent Range 97+%
Molecular Formula C9H14OSi
Linear Formula C6H5OSi(CH3)3
Formula Weight 166.29g/mol
MDL Number MFCD00053666
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.