missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenoxyacetic acid, 98+%, ACROS Organics™

Manufacturer:  Acros Organics 169871000

 View more versions of this product

Catalog No. AC169871000



Specifications

Specifications

Phenoxyacetic acid
Authentic
0.1% max.
285.0°C
98.0°C to 101.0°C
122-59-8
C8H8O3
phenoxyacetic acid, phenoxyethanoic acid, acetic acid, phenoxy, glycolic acid phenyl ether, phenoxy acetic acid, glycol acid phenyl ether, o-phenylglycolic acid, phenoxyacetate, glycollic acid phenyl ether, acide phenoxyacetique
LCPDWSOZIOUXRV-UHFFFAOYSA-N
2-phenoxyacetic acid
19188
152.15
≥98%
165°C
Plastic bottle
0.5% max.
White
100g
98+%
C6H5OCH2CO2H
Solubility in water: soluble. Other solubilities: soluble in glacial acetic acid, soluble in ether, alcohol and carbon disulfide
C1=CC=C(C=C1)OCC(=O)O
152.149
CHEBI:8075
Granular Powder, Granules, or Prills

Chemical Identifiers

122-59-8
152.149
phenoxyacetic acid, phenoxyethanoic acid, acetic acid, phenoxy, glycolic acid phenyl ether, phenoxy acetic acid, glycol acid phenyl ether, o-phenylglycolic acid, phenoxyacetate, glycollic acid phenyl ether, acide phenoxyacetique
CHEBI:8075
C1=CC=C(C=C1)OCC(=O)O
C8H8O3
LCPDWSOZIOUXRV-UHFFFAOYSA-N
19188
2-phenoxyacetic acid
Safety and Handling

Safety and Handling

Warning

SDS Documents