Please login to your online account to display your discounted pricing

Phenothiazine-10-carbonyl chloride, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Phenothiazine-10-carbonyl chloride
Name Note 98%
CAS 18956-87-1
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C13H8ClNOS
Formula Weight 261.71
MDL Number MFCD00044216
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC167210050

 
fisher scientific acros
167210050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 261.71
Physical Form Crystalline Powder
Quantity 5g
Color Light green or yellow-brown
Melting Point 168° to 172°C
EINECS Number 242-699-7
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Name Note 98%
Assay Percent Range 98%
MDL Number MFCD00044216
CAS 18956-87-1
Chemical Name or Material Phenothiazine-10-carbonyl chloride
Molecular Formula C13H8ClNOS
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.