missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Phenolphthalin 98.0+%, TCI America™
$109.49 - $776.41
Chemical Identifiers
CAS | 81-90-3 |
---|---|
Molecular Formula | C20H16O4 |
Molecular Weight (g/mol) | 320.344 |
MDL Number | MFCD00036135 |
InChI Key | FFFPYJTVNSSLBQ-UHFFFAOYSA-N |
PubChem CID | 66494 |
ChEBI | CHEBI:34915 |
IUPAC Name | 2-[bis(4-hydroxyphenyl)methyl]benzoic acid |
SMILES | C1=CC=C(C(=C1)C(C2=CC=C(C=C2)O)C3=CC=C(C=C3)O)C(=O)O |
Chemical Identifiers
81-90-3 | |
320.344 | |
FFFPYJTVNSSLBQ-UHFFFAOYSA-N | |
CHEBI:34915 | |
C1=CC=C(C(=C1)C(C2=CC=C(C=C2)O)C3=CC=C(C=C3)O)C(=O)O |
C20H16O4 | |
MFCD00036135 | |
66494 | |
2-[bis(4-hydroxyphenyl)methyl]benzoic acid |
Specifications
235°C | |
C20H16O4 | |
FFFPYJTVNSSLBQ-UHFFFAOYSA-N | |
2-[bis(4-hydroxyphenyl)methyl]benzoic acid | |
66494 | |
320.34 | |
White | |
Crystal/Powder |
81-90-3 | |
MFCD00036135 | |
C1=CC=C(C(=C1)C(C2=CC=C(C=C2)O)C3=CC=C(C=C3)O)C(=O)O | |
320.344 | |
CHEBI:34915 | |
≥98.0% (HPLC,T) | |
25 g | |
Phenolphthalin [for redox indicator] |
Safety and Handling
EINECSNumber : (4)-0263
TSCA : Yes