Please login to your online account to display your discounted pricing

Phenolphthalein TS/RS, 1% (w/v) Alcoholic, Ricca Chemical


Quantity 250mL

Manufacturer: ricca chemical company  RIR5622000250A

 View more versions of this product

Catalog No. R5622000250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Quantity 250mL
Chemical Name or Material Phenolphthalein
CAS Max % 6.0
CAS 7732-18-5 (Water, Deionized)
Name Note Test Solution/Reagent Solution
Physical Form ear, colorless to slight pink liquid
CAS 64-17-5
CAS Min % 94.0
CAS Max % 96.0
CAS 77-09-8
CAS Min % 1.21
CAS Max % 1.26
CAS 67-56-1
CAS Min % 4.0

Specifications

Chemical Name or Material Phenolphthalein
Name Note Test Solution/Reagent Solution
CAS 64-17-5
CAS Min % 94.0
CAS Max % 96.0
CAS 77-09-8
CAS Min % 1.21
CAS Max % 1.26
CAS 67-56-1
CAS Min % 4.0
CAS Max % 6.0
CAS 7732-18-5 (Water, Deionized)
Physical Form ear, colorless to slight pink liquid