missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenolphthalein TS/RS, 1% (w/v) Alcoholic, Ricca Chemical

$73.58 - $73.58

Chemical Identifiers

CAS 64-17-5,77-09-8,67-56-1,7732-18-5 (Water, Deionized)
CAS Min % 94.0,1.21,4.0
CAS Max % 96.0,1.26,6.0
Physical Form Liquid
Chemical Name or Material Phenolphthalein, Test Solution/Reagent Solution
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
5622-1 SDS Ricca Chemical Company
RI56221
Poly natural 4L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
5622-32 Ricca Chemical Company
RI562232
Poly natural 1L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
R5622000250 Ricca Chemical Company
RIR5622000250A
250mL Each for $73.58
Specifications Description & Specifications

Chemical Identifiers

64-17-5,77-09-8,67-56-1,7732-18-5 (Water, Deionized)
96.0,1.26,6.0
Phenolphthalein, Test Solution/Reagent Solution
94.0,1.21,4.0
Liquid

Specifications

64-17-5,77-09-8,67-56-1,7732-18-5 (Water, Deionized)
96.0,1.26,6.0
Poly natural
Alcoholic Solution
4L
94.0,1.21,4.0
Liquid
1% (w/v)
Clear colorless to slight pink
Phenolphthalein, Test Solution/Reagent Solution