Please login to your online account to display your discounted pricing

Phenolphthalein Solution R, 0.1% (w/v) in 80% (v/v) Alcohol, Ricca Chemical

$84.85 - $226.70

Chemical Identifiers

Chemical Name or Material Phenolphthalein Solution
Name Note Reagent
CAS 64-17-5
CAS Min % 75.0
CAS Max % 77.0
CAS 7732-18-5
CAS Min % 19.0
CAS Max % 21.0
CAS 77-09-8
CAS Min % 0.12
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

5598-32

 
ricca chemical company
RI559832
Poly natural 1L Each for $84.85

55988

  SDS
ricca chemical company
RI55988
Poly natural 250mL Each for $226.70
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless to slight pink
Quantity 1L
Concentration or Composition Notes 0.1% (w/v)
Solvent or Matrix 80% (v/v) Alcohol
Chemical Name or Material Phenolphthalein Solution
CAS Max % 0.12
CAS 67-56-1
CAS Min % 3.0
CAS Max % 5.0
Name Note Reagent
CAS 64-17-5
CAS Min % 75.0
Packaging Poly natural
CAS Max % 77.0
CAS 7732-18-5
CAS Min % 19.0
CAS Max % 21.0
CAS 77-09-8
CAS Min % 0.12