missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell Fluka™ Phenolphthalein solution, Indicator, Reag. Ph. Eur., 1% in ethanol, Honeywell Fluka™

Indicator, Reag. Ph. Eur., 1% in ethanol

Manufacturer:  Honeywell Fluka™ 346076X500ML

 View more versions of this product

Catalog No. 60046379



Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Specifications

Phenolphthalein solution
77-09-8
C20H14O4
284423
C1=CC=C2C(=C1)C(=O)OC2(C3=CC=C(C=C3)O)C4=CC=C(C=C4)O
318.328
CHEBI:34914
Poly Bottle
Indicator, Reag. Ph. Eur., 1% in ethanol
C20H14O4
MFCD00005913
KJFMBFZCATUALV-UHFFFAOYSA-N
3,3-bis(4-hydroxyphenyl)-2-benzofuran-1-one
4764
Reagent Ph. Eur.
6 x 500mL

Chemical Identifiers

77-09-8
318.328
KJFMBFZCATUALV-UHFFFAOYSA-N
CHEBI:34914
C1=CC=C2C(=C1)C(=O)OC2(C3=CC=C(C=C3)O)C4=CC=C(C=C4)O
C20H14O4
MFCD00005913
4764
3,3-bis(4-hydroxyphenyl)-2-benzofuran-1-one
Safety and Handling

Safety and Handling

ShelfLife : 1440 days from date of manufacture

Documents