Please login to your online account to display your discounted pricing

Phenolphthalein Indicator, 1% (w/v) in 70% (v/v) Alcohol, Neutralized, Ricca Chemical

$67.85 - $67.85

Chemical Identifiers

Chemical Name or Material Phenolphthalein Indicator
CAS 7732-18-5
CAS Min % 29.0
CAS Max % 31.0
CAS 64-17-5
CAS Min % 65.0
CAS Max % 69.0
CAS 77-09-8
CAS Min % 0.5
CAS Max % 1.5
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

5618-16

  SDS
ricca chemical company
RI561816
Poly natural 500mL Each for $67.85

5618-32

 
ricca chemical company
RI561832
Poly natural 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

5618-4

 
ricca chemical company
RI56184
Poly natural 120mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless to slight pink
Quantity 500mL
Treatment(s) Neutralized
Concentration or Composition Notes 1% (w/v)
Solvent or Matrix 70% (v/v) Alcohol
Chemical Name or Material Phenolphthalein Indicator
CAS 67-56-1
CAS 7732-18-5
CAS 77-09-8
CAS Max % 1.5
CAS Min % 2.0
CAS Max % 4.0
CAS Min % 29.0
Packaging Poly natural
CAS Max % 31.0
CAS 64-17-5
CAS Min % 65.0
CAS Max % 69.0
CAS Min % 0.5