Please login to your online account to display your discounted pricing

Phenolphthalein Disodium Salt 95%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: fisher scientific acros  417211000

 View more versions of this product

Catalog No. AC417211000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder
Purity Grade Extra Pure
Formula Weight 362.28
Quantity 100g
Color Dark purple to dark brown
Merck Index 12, 7393
TSCA TSCA
EINECS Number 208-254-6
Loss on Drying 9% max. (1 g, 105°C, 3 hrs)
Packaging Glass bottle
Infrared Spectrum Authentic
Name Note 95%
UV_Vis 94% min. (on dry substance)
Chemical Name or Material Phenolphthalein disodium salt
CAS 518-51-4
Molecular Formula C20H12Na2O4
Assay Percent Range 95%
MDL Number MFCD00051414

Specifications

Chemical Name or Material Phenolphthalein disodium salt
Name Note 95%
CAS 518-51-4
Purity Grade Extra Pure
Assay Percent Range 95%
Molecular Formula C20H12Na2O4
Formula Weight 362.28
MDL Number MFCD00051414