Please login to your online account to display your discounted pricing

Phenolphthalein Disodium Salt 95%, ACROS Organics™

$317.71 - $317.71

Chemical Identifiers

Chemical Name or Material Phenolphthalein disodium salt
Name Note 95%
CAS 518-51-4
Purity Grade Extra Pure
Assay Percent Range 95%
Molecular Formula C20H12Na2O4
Formula Weight 362.28
MDL Number MFCD00051414
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC417211000

  SDS
fisher scientific acros
417211000
Glass bottle 100g Each for $317.71
Description & Specifications

Specifications

Physical Form Powder
Purity Grade Extra Pure
Formula Weight 362.28
Quantity 100g
Color Dark purple to dark brown
Merck Index 12, 7393
TSCA TSCA
EINECS Number 208-254-6
Chemical Name or Material Phenolphthalein disodium salt
Name Note 95%
Assay Percent Range 95%
Molecular Formula C20H12Na2O4
MDL Number MFCD00051414
CAS 518-51-4
Infrared Spectrum Authentic
Loss on Drying 9% max. (1 g, 105°C, 3 hrs)
Packaging Glass bottle
UV_Vis 94% min. (on dry substance)