Please login to your online account to display your discounted pricing

Phenolphthalein, ACS Grade, LabChem™

$65.23 - $65.23

Chemical Identifiers

Chemical Name or Material Phenolphthalein
CAS 77-09-8
CAS Max % 100
Purity Grade ACS Grade
Linear Formula C20H14O4
Formula Weight 318.32
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC181989

 
lab chem
LC181989
100g Each for $65.23
Description & Specifications

Specifications

Formula Weight 318.32
Chemical Name or Material Phenolphthalein
Quantity 100g
EINECS Number 201-004-7
Purity Grade ACS Grade
Physical Form Powder
Linear Formula C20H14O4
CAS 77-09-8
Identification Passes Test
Health Hazard 1 Warning
Packaging Poly Bottle
CAS Max % 100
Health Hazard 2 GHS H Statement
Suspected of causing cancer.
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear protective gloves/eye protection/face protection.
If exposed or concerned: Get medical attention.
Store locked up.
Dispose of contents/container in accordance with local, state and federal regulations.
Solubility Soluble in water
Specific Gravity 1277kg/m3
Recommended Storage Room Temperature
Decomposition Information Carbon dioxide; Carbon monoxide
Density 1,277kg/m3
Boiling Point 450°C
Melting Point 262°C
Color White
Safety and Handling

Warning

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear protective gloves/eye protection/face protection.
If exposed or concerned: Get medical attention.
Store locked up.
Dispose of contents/container in accordance with local, state and federal regulations.

GHS H Statement
Suspected of causing cancer.

Room Temperature