missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Phenolphthalein, 2% soln. in 95% ethanol

pH indicator

$74.16 - $74.16

Chemical Identifiers

CAS 77-09-8
Molecular Formula C20H14O4
Molecular Weight (g/mol) 318.328
MDL Number MFCD00005913
InChI Key KJFMBFZCATUALV-UHFFFAOYSA-N
Synonym phenolphthalein, phthalimetten, euchessina, phthalin, espotabs, phenolax, purgophen, koprol, laxogen, trilax
PubChem CID 4764
ChEBI CHEBI:34914
IUPAC Name 3,3-bis(4-hydroxyphenyl)-2-benzofuran-1-one
SMILES C1=CC=C2C(=C1)C(=O)OC2(C3=CC=C(C=C3)O)C4=CC=C(C=C4)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ61785AU View Documents Alfa Aesar™
J61785AU
1000mL Each for $74.16
 
Specifications

Chemical Identifiers

77-09-8
318.328
KJFMBFZCATUALV-UHFFFAOYSA-N
4764
3,3-bis(4-hydroxyphenyl)-2-benzofuran-1-one
C20H14O4
MFCD00005913
phenolphthalein, phthalimetten, euchessina, phthalin, espotabs, phenolax, purgophen, koprol, laxogen, trilax
CHEBI:34914
C1=CC=C2C(=C1)C(=O)OC2(C3=CC=C(C=C3)O)C4=CC=C(C=C4)O

Specifications

77-09-8
MFCD00005913
2% soln. in 95% ethanol
phenolphthalein, phthalimetten, euchessina, phthalin, espotabs, phenolax, purgophen, koprol, laxogen, trilax
C1=CC=C2C(=C1)C(=O)OC2(C3=CC=C(C=C3)O)C4=CC=C(C=C4)O
318.328
CHEBI:34914
Liquid
Alcohol-like
1000mL
Phenolphthalein
C20H14O4
UN1987
14,7243
KJFMBFZCATUALV-UHFFFAOYSA-N
3,3-bis(4-hydroxyphenyl)-2-benzofuran-1-one
4764
318.33
95% ethanol
Colorless
17°C (63°F)
Safety and Handling

Safety and Handling

GHS H Statement
H225-H350-H341-H361-H371
Highly flammable liquid and vapor.
May cause cancer.
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.
May cause damage to organs.

GHS P Statement
P210-P260-P280-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALCOHOLS, N.O.S.

TSCA : TSCA