Please login to your online account to display your discounted pricing

Alfa Aesar™ Phenolphthalein, 1% w/v in alcohol

Chemical Identifiers

Chemical Name or Material Phenolphthalein
CAS 77-09-8
Formula Weight 318.33
MDL Number MFCD00005913
UN Number UN1987
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA38704AP

  SDS
alfa aesar™
38704AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 318.33
Health Hazard 3 GHS P Statement
P210-P260-P280-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
TSCA TSCA
UN Number UN1987
MDL Number MFCD00005913
Concentration 1% w/v in alcohol
Color Colorless
Quantity 500mL
Physical Form Liquid
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H350-H341-H361-H371
Highly flammable liquid and vapor.
May cause cancer.
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.
May cause damage to organs.
Chemical Name or Material Phenolphthalein
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALCOHOLS, N.O.S.
CAS 77-09-8
Odor Alcohol-like
Flash Point 11° (52°F)
Safety and Handling

Danger

GHS P Statement
P210-P260-P280-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.

GHS H Statement
H225-H350-H341-H361-H371
Highly flammable liquid and vapor.
May cause cancer.
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.
May cause damage to organs.