missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Phenolphthalein, 1% (w/v) in 60% (v/v) Isopropyl Alcohol, Neutralized, Ricca Chemical
Supplier: Ricca Chemical Company R56256001A
Specifications
Phenolphthalein | |
67-63-0,77-09-8 | |
Natural Poly Bottle | |
Colorless | |
Liquid |
1% (w/v) in 60% (v/v) Isopropyl Alcohol, Neutralized | |
dihydrogen oxide, dihydrogen monoxide | |
Aqueous Solution | |
1 L |