Please login to your online account to display your discounted pricing

Phenolphthalein, 1% (w/v) in 50% (v/v) Isopropyl Alcohol, Neutralized, Ricca Chemical

$57.64 - $57.64

Chemical Identifiers

Chemical Name or Material Phenolphthalein
CAS 7732-18-5
CAS Min % 49.0
CAS Max % 51.0
CAS 67-63-0
CAS Min % 49.0
CAS Max % 51.0
CAS 77-09-8
CAS Min % 0.5
CAS Max % 1.5
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

5625-16

 
ricca chemical company
RI562516
Poly natural 500mL Each for $57.64

R56250004A

 
ricca chemical company
RIR56250004A
Plastic Bottle 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R562500060A

 
ricca chemical company
RIR562500060A
Plastic Bottle 60mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless to slight pink
Quantity 500mL
Concentration or Composition Notes 1% (w/v)
Solvent or Matrix 50% (v/v) Isopropyl Alcohol
Treatment(s) Neutralized
CAS 7732-18-5
Chemical Name or Material Phenolphthalein
CAS 77-09-8
CAS 67-63-0
CAS Min % 0.5
CAS Max % 1.5
CAS Max % 51.0
CAS Max % 51.0
CAS Min % 49.0
CAS Min % 49.0
Packaging Poly natural