missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenolphthalein, 1% In 60% Isopropanol, pH 8.3 to 10.0 Colorless to Pink, Certified, LabChem™

Manufacturer:  Lab Chem LC182107

 View more versions of this product

Catalog No. LC182107


 

Specifications Description & Specifications

Specifications

Phenolphthalein
67-63-0,7732-18-5,77-09-8
Certified Grade
C20H14O4
UN1219
Poly Bottle
Carbon dioxide; Carbon monoxide; Fume; May release flammable gases
125mL
1% In 60% Isopropanol, pH 8.3 to 10.0 Colorless to Pink
60,39,1
Liquid
318.32
Passes Test
Soluble in water
Clear, colorless to pink

Chemical Identifiers

67-63-0,7732-18-5,77-09-8
C20H14O4
UN1219
Certified Grade
60,39,1
318.32
Phenolphthalein, 1% In 60% Isopropanol, pH 8.3 to 10.0 Colorless to Pink

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of causing cancer.

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, ventilating, lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, vapors, spray.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
If exposed or concerned: Get medical advice/attention.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
In case of fire: Use dry chemical powder, alcohol-resistant foam, carbon dioxide to extinguish.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.

Danger

EINECSNumber : 200-661-7

Recommended Storage : Room Temperature

Promotions

Promotions

  • Buy one of the LabChem™ Chemicals listed on this page and get another chemical at no additional cost. This is an NLSU exclusive offer. You must apply and qualify for the program to be eligible.

    06/30/18