Please login to your online account to display your discounted pricing

Phenolphthalein, 1% in 95% Ethanol, pH 8.3 to 10.0 Colorless to Pink, Certified, LabChem™

$20.66 - $169.29

Chemical Identifiers

Chemical Name or Material Phenolphthalein
CAS 64-17-5
CAS Max % 89.1
CAS 67-63-0
CAS Max % 4.95
CAS 67-56-1
CAS Max % 4.95
CAS 77-09-8
CAS Max % 1
Name Note 1% in 95% Ethanol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC182201

 
lab chem
LC182201
500mL Each for $32.00

LC182202

 
lab chem
LC182202
1L Each for $54.63

LC182204

 
lab chem
LC182204
4L Each for $169.29

LC182207

 
lab chem
LC182207
125mL Each for $20.66
Description & Specifications

Specifications

Purity Grade Certified Grade
pH 8.3 to 10.0
Formula Weight 318.32
Health Hazard 2 GHS H Statement
Highly flammable liquid and vapor.
Harmful if swallowed.
Causes skin and serious eye irritation.
May cause respiratory irritation.
Suspected of causing cancer.
Suspected of damaging fertility or the unborn chi
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep container tightly closed.<
Health Hazard 1 Danger
Chemical Name or Material Phenolphthalein
CAS 77-09-8
CAS 67-63-0
Decomposition Information fume; Carbon monoxide; Carbon dioxide; May release flammable gases
CAS Max % 4.95
Physical Form Liquid
CAS 64-17-5
Packaging Poly Bottle
CAS Max % 4.95
CAS Max % 1
Name Note 1% in 95% Ethanol
CAS Max % 89.1
Identification Passes Test
Recommended Storage Room Temperature
Specific Gravity 0.95g/mL
Linear Formula C20H14O4
Color clear, colorless to pink solution
Quantity 500mL
UN Number UN1170
Solubility Soluble in water
Density 0.95g/mL
CAS 67-56-1
Safety and Handling

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep container tightly closed.<

GHS H Statement
Highly flammable liquid and vapor.
Harmful if swallowed.
Causes skin and serious eye irritation.
May cause respiratory irritation.
Suspected of causing cancer.
Suspected of damaging fertility or the unborn chi

Room Temperature