missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Phenolphthalein, 1% in 95% Ethanol, pH 8.3 to 10.0 Colorless to Pink, Certified, LabChem™
Supplier: LabChem LC182204
Specifications
Phenolphthalein | |
89.1,4.95,4.95,1 | |
C20H14O4 | |
Passes Test | |
Certified | |
0.95g/mL | |
0.95g/mL | |
4 L |
1% in 95% Ethanol, pH 8.3 to 10.0 Colorless to Pink | |
67-63-0,67-56-1,77-09-8 | |
Soluble in water | |
318.32 | |
Poly Bottle | |
Fume; Carbon monoxide; Carbon dioxide; May release flammable gases | |
Colorless to Pink | |
Liquid |