Please login to your online account to display your discounted pricing

Phenolphthalein, 0.5% In 50% Ethanol, pH 8.3 to 10.0 Colorless to Pink, Certified, LabChem™


Quantity 125mL

Manufacturer: lab chem  LC182007

 View more versions of this product

Catalog No. LC182007

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 318.32
Purity Grade Certified Grade
Linear Formula C20H14O4
CAS 67-56-1
Health Hazard 1 Danger
Specific Gravity 0.95g/mL
CAS Max % 0.5
Density 0.95g/mL
Identification Passes Test
Decomposition Information Carbon monoxide; Carbon dioxide
CAS 64-17-5
CAS 7732-18-5
CAS Max % 2.4
Physical Form Liquid
Health Hazard 2 GHS H Statement
Flammable liquid and vapor.
May cause cancer.
Suspected of damaging the unborn child (oral).
Causes damage to organs (central nervous system, optic nerve) (oral, dermal).
UN Number UN1170
Solubility Soluble in water
CAS 67-63-0
Name Note 0.5% In 50% Ethanol, pH 8.3 to 10.0 Colorless to Pink
Recommended Storage Room Temperature
CAS Max % 2.5
CAS Max % 44.8
Chemical Name or Material Phenolphthalein
Color Clear, colorless to pink
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep away from heat, sparks, open flames, hot surfaces.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, ventilating, lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, vapors, spray.
Do not eat, drink or smoke when using this product.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
If exposed or concerned: Get medical advice/attention.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
In case of fire: Use carbon dioxide (CO2), powder, alcohol-resistant foam to extinguish.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.
CAS 77-09-8
Packaging Poly Bottle
Quantity 125mL
CAS Max % 49.8

Specifications

Chemical Name or Material Phenolphthalein
CAS 7732-18-5
CAS Max % 49.8
CAS 64-17-5
CAS Max % 44.8
CAS 67-63-0
CAS Max % 2.5
CAS 67-56-1
CAS Max % 2.4
CAS 77-09-8
CAS Max % 0.5
Name Note 0.5% In 50% Ethanol, pH 8.3 to 10.0 Colorless to Pink
Purity Grade Certified Grade
Linear Formula C20H14O4
Formula Weight 318.32
UN Number UN1170
Accessories

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep away from heat, sparks, open flames, hot surfaces.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, ventilating, lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, vapors, spray.
Do not eat, drink or smoke when using this product.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
If exposed or concerned: Get medical advice/attention.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
In case of fire: Use carbon dioxide (CO2), powder, alcohol-resistant foam to extinguish.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Flammable liquid and vapor.
May cause cancer.
Suspected of damaging the unborn child (oral).
Causes damage to organs (central nervous system, optic nerve) (oral, dermal).

Room Temperature