missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenolphthalein, 0.08% in Methanol, for Organic Acids, Certified, LabChem™

Manufacturer:  LabChem LC182307

 View more versions of this product

Catalog No. LC182307



Specifications

Specifications

Phenolphthalein
77-09-8,67-56-1
C20H14O4
UN1993
KJFMBFZCATUALV-UHFFFAOYSA-N
3,3-bis(4-hydroxyphenyl)-2-benzofuran-1-one
4764
318.32
Certified
Soluble in water
Fume; Carbon monoxide; Carbon dioxide; May release flammable gases
65°C
Colorless
11°C
0.08% in Methanol, for Organic Acids
99.92,0.08
C20H14O4
Passes Test
C1=CC=C2C(=C1)C(=O)OC2(C3=CC=C(C=C3)O)C4=CC=C(C=C4)O
318.328
CHEBI:34914
Liquid
Poly Bottle
0.792g/mL
0.792g/mL
-98°C
125mL

Chemical Identifiers

77-09-8
318.328
4764
3,3-bis(4-hydroxyphenyl)-2-benzofuran-1-one
C20H14O4
KJFMBFZCATUALV-UHFFFAOYSA-N
CHEBI:34914
C1=CC=C2C(=C1)C(=O)OC2(C3=CC=C(C=C3)O)C4=CC=C(C=C4)O
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.
Causes damage to organs (central nervous system, optic nerve) (oral, dermal).

GHS P Statement
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, ventilating, lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, vapors, spray.
Do not eat, drink or smoke when using this product.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Store locked up in a cool, well-ventilated place.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
In case of fire: Use carbon dioxide (CO2), powder, alcohol-resistant foam to extinguish.
Dispose of contents/container to comply with local, state and federal regulations.

Danger

Recommended Storage : Room Temperature

Documents
Promotions

Promotions