Please login to your online account to display your discounted pricing

Phenol USP Crystal, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Phenol
CAS 108-95-2
Purity Grade USP
Physical Form Crystalline
Molecular Formula C6H5OH
Formula Weight 94.11
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK0605212

 
avantor performance
060506
Poly Coated 2.5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MK0605500

 
avantor performance
060504
Glass 12 x 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Phenol
Quantity 2.5kg
CAS 108-95-2
Purity Grade USP
Packaging Poly Coated
Physical Form Crystalline
Molecular Formula C6H5OH
Formula Weight 94.11