Please login to your online account to display your discounted pricing

Phenol USP Crystal, Macron Fine Chemicals™


macron

Quantity 2.5kg
Packaging Poly Coated

Manufacturer: avantor performance  060506

 View more versions of this product

Catalog No. MK0605212

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Phenol
Quantity 2.5kg
CAS 108-95-2
Purity Grade USP
Packaging Poly Coated
Physical Form Crystalline
Molecular Formula C6H5OH
Formula Weight 94.11

Specifications

Chemical Name or Material Phenol
CAS 108-95-2
Purity Grade USP
Physical Form Crystalline
Molecular Formula C6H5OH
Formula Weight 94.11