Please login to your online account to display your discounted pricing

Phenol, Ultra Pure (MP Biomedicals80067-), MP Biomedicals


Quantity 100g

Manufacturer: mp biomedicals  800672

 View more versions of this product

Catalog No. ICN800672

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 100g
Purity Grade UltraPure
CAS 108-95-2
Chemical Name or Material Phenol

Specifications

Chemical Name or Material Phenol
CAS 108-95-2
Purity Grade UltraPure