Please login to your online account to display your discounted pricing

Phenol, Ultra Pure (MP Biomedicals80067-), MP Biomedicals

$70.77 - $227.67

Chemical Identifiers

Chemical Name or Material Phenol
CAS 108-95-2
Purity Grade UltraPure
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN800673

 
mp biomedicals
800673
1kg Each for $227.67

ICN800672

 
mp biomedicals
800672
100g Each for $70.77
Description & Specifications

Specifications

Quantity 1kg
Purity Grade UltraPure
Chemical Name or Material Phenol
CAS 108-95-2