missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Phenol 99.5+%, TCI America™
$34.84 - $68.02
Chemical Identifiers
CAS | 108-95-2 |
---|---|
Molecular Formula | C6H6O |
Molecular Weight (g/mol) | 94.11 |
MDL Number | MFCD00002143 |
InChI Key | ISWSIDIOOBJBQZ-UHFFFAOYSA-N |
PubChem CID | 996 |
ChEBI | CHEBI:15882 |
IUPAC Name | phenol |
SMILES | OC1=CC=CC=C1 |
Chemical Identifiers
108-95-2 | |
94.11 | |
ISWSIDIOOBJBQZ-UHFFFAOYSA-N | |
CHEBI:15882 | |
OC1=CC=CC=C1 |
C6H6O | |
MFCD00002143 | |
996 | |
phenol |
Specifications
42°C | |
108-95-2 | |
MFCD00002143 | |
ISWSIDIOOBJBQZ-UHFFFAOYSA-N | |
phenol | |
996 | |
94.11 | |
Red/White | |
25 g | |
Phenol |
181°C | |
C6H6O | |
1671 | |
OC1=CC=CC=C1 | |
94.11 | |
CHEBI:15882 | |
≥99.5% (GC) | |
6.0 (H2O soln.) | |
Crystal/Lump |
Safety and Handling
EINECSNumber : (3)-0481
RTECSNumber : SJ3325000
TSCA : Yes