Please login to your online account to display your discounted pricing

Phenol, Saturated (pH 4.3, Liq.), Fisher BioReagents


Quantity 100mL
Packaging Coated Amber Glass/Poison Pack

Manufacturer: fisher scientific company  BP1751I100

 View more versions of this product

Catalog No. BP1751I-100

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Quantity 100mL
DOT Information DOT Class 6.1, : Poison
Physical Form Liquid
pH 4.3
Density 1.05g/cm³
Identification Pass Test
Assay Percent Range ≥99.0
Health Hazard 2 DANGER!
Health Hazard 3 Emergency Overview
Combustible liquid. Causes burns by all exposure routes. May cause central nervous system effects. Possible risks of irreversible effects. Danger of serious damage to health by prolonged exposure. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Immediate medical attention is required. Move to fresh air.

NFPA

Health:4
Flammability:2
Instability:0
CAS 108-95-2
ChemAlert Storage Symbol White
Packaging Coated Amber Glass/Poison Pack
Optical Density 330nm (0.2 AU max.), 405nm (0.1 AU), 510nm (0.1nm)
Recommended Storage 4°C
Chemical Name or Material Phenol, Saturated

Specifications

Chemical Name or Material Phenol, Saturated
CAS 108-95-2
Physical Form Liquid
Assay ≥99.0%
Assay Percent Range ≥99.0
Accessories
 

Emergency Overview
Combustible liquid. Causes burns by all exposure routes. May cause central nervous system effects. Possible risks of irreversible effects. Danger of serious damage to health by prolonged exposure. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Immediate medical attention is required. Move to fresh air.

NFPA

Health:4
Flammability:2
Instability:0

DANGER!

4°C

SDS & Safety Documents