missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenol, Saturated (pH 4.3, Liq.), Fisher BioReagents

$177.71 - $302.63

Chemical Identifiers

CAS 108-95-2
Molecular Formula C6H6O
Molecular Weight (g/mol) 94.113
InChI Key ISWSIDIOOBJBQZ-UHFFFAOYSA-N
Synonym carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol
PubChem CID 996
ChEBI CHEBI:15882
IUPAC Name phenol
SMILES C1=CC=C(C=C1)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
BP1751I-400 SDS View Documents Fisher BioReagents
BP1751I400
400mL Coated Amber Glass/Poison Pack Each for $302.63
BP1751I-100 SDS View Documents Fisher BioReagents
BP1751I100
100mL Coated Amber Glass/Poison Pack Each for $177.71
Description

Description

Phenol saturated with Tris buffer is used for purifying RNA.
Specifications

Chemical Identifiers

108-95-2
94.113
carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol
CHEBI:15882
C1=CC=C(C=C1)O
C6H6O
ISWSIDIOOBJBQZ-UHFFFAOYSA-N
996
phenol

Specifications

108-95-2
C6H6O
carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol
C1=CC=C(C=C1)O
94.113
CHEBI:15882
≥99.0%
Coated Amber Glass/Poison Pack
4.3
Phenol, Saturated
≥99.0
1.05g/cm³
ISWSIDIOOBJBQZ-UHFFFAOYSA-N
phenol
996
Liquid
Pass Test
330nm (0.2 AU max.), 405nm (0.1 AU), 510nm (0.1nm)
400mL
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Combustible liquid. Causes burns by all exposure routes. May cause central nervous system effects. Possible risks of irreversible effects. Danger of serious damage to health by prolonged exposure. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Immediate medical attention is required. Move to fresh air.

NFPA

Health:4
Flammability:2
Instability:0

DOTInformation : DOT Class 6.1, : Poison

Recommended Storage : 4°C