Please login to your online account to display your discounted pricing

Phenol Red, Sodium Salt, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 1kg
Packaging Wide Mouth Amber Glass Bottle

Manufacturer: avantor performance  T26508

 View more versions of this product

Catalog No. 02003631

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 376.37
Physical Form Powder
Molecular Formula NaOSO2C6H4C(C6H4-4-OH):C6H4-4-(:O)
CAS 34487-61-1
Quantity 1kg
Packaging Wide Mouth Amber Glass Bottle
Chemical Name or Material Phenol Red, Sodium Salt

Specifications

Chemical Name or Material Phenol Red, Sodium Salt
CAS 34487-61-1
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula NaOSO2C6H4C(C6H4-4-OH):C6H4-4-(:O)
Formula Weight 376.37