Please login to your online account to display your discounted pricing

Phenol red sodium salt, ACS reagent grade, MP Biomedicals™


Quantity 5g

Manufacturer: mp biomedicals  0215255505

 View more versions of this product

Catalog No. ICN15255505

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical State Solid
Quantity 5g
Chemical Name or Material Phenol red sodium salt
CAS 34487-61-1
Purity Grade ACS Reagent
Molecular Formula C19H13O5SNa
Formula Weight 376.4g/mol
Synonym Phenolsulfonephthalein sodium salt
Recommended Storage Store at room temperature.
EINECS Number 252-057-8

Specifications

Chemical Name or Material Phenol red sodium salt
CAS 34487-61-1
Purity Grade ACS Reagent
Molecular Formula C19H13O5SNa
Formula Weight 376.4g/mol
Synonym Phenolsulfonephthalein sodium salt
Accessories

Store at room temperature.