Please login to your online account to display your discounted pricing

Phenol red sodium salt, ACS reagent grade, MP Biomedicals™

$53.40 - $53.40

Chemical Identifiers

Chemical Name or Material Phenol red sodium salt
CAS 34487-61-1
Purity Grade ACS Reagent Grade
Molecular Formula C19H13O5SNa
Formula Weight 376.4
Synonym Phenolsulfonephthalein sodium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN15255510

 
mp biomedicals
15255510
10g Each for $53.40
Description & Specifications

Specifications

Purity Grade ACS Reagent Grade
Formula Weight 376.4
Quantity 10g
EINECS Number 252-057-8
Physical State Solid
CAS 34487-61-1
Chemical Name or Material Phenol red sodium salt
Recommended Storage Store at room temperature.
Synonym Phenolsulfonephthalein sodium salt
Molecular Formula C19H13O5SNa