Please login to your online account to display your discounted pricing

Phenol Red Sodium Salt, ACS Grade, LabChem™

$71.69 - $71.69

Chemical Identifiers

Chemical Name or Material Phenol Red Sodium Salt
CAS 34487-61-1
CAS Max % 100
Purity Grade ACS Grade
Linear Formula C19H13O5SNa
Formula Weight 376.36
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC182407

 
lab chem
LC182407
5g Each for $71.69
Description & Specifications

Specifications

Formula Weight 376.36
Health Hazard 3 GHS P Statement
Avoid breathing dust.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.
CAS 34487-61-1
Health Hazard 1 Warning
Physical Form Powder
Melting Point 285°C
CAS Max % 100
EINECS Number 252-057-8
Recommended Storage Room Temperature
Purity Grade ACS Grade
Identification Passes Test
Quantity 5g
Decomposition Information Sulfur compounds; Carbon monoxide; Carbon dioxide
Color Dark red
Chemical Name or Material Phenol Red Sodium Salt
Linear Formula C19H13O5SNa
Health Hazard 2 GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Solubility Soluble in water
Packaging Amber Glass
Safety and Handling

Warning

GHS P Statement
Avoid breathing dust.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Room Temperature