Please login to your online account to display your discounted pricing

Phenol red, MP Biomedicals™


Phenol Red Sodium Salt is used as a pH indicator, it is used widely in culture media to identify changes from neutral to acidic pH values

Quantity 5g

Manufacturer: mp biomedicals  10260405

 View more versions of this product

Catalog No. ICN10260405

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Shiny Powder
Formula Weight 376.4
Color Dark red
Quantity 5g
EINECS Number 252-057-8
Chemical Name or Material Phenol Red Sodium Salt
Molecular Formula C19H13O5SNa
MDL Number MFCD00066901
CAS 34487-61-1
Synonym Phenolsulfonphthalein

Specifications

Chemical Name or Material Phenol Red Sodium Salt
CAS 34487-61-1
Physical Form Shiny Powder
Molecular Formula C19H13O5SNa
Formula Weight 376.4
MDL Number MFCD00066901
Synonym Phenolsulfonphthalein