missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenol red, MP Biomedicals™

Phenol Red Sodium Salt is used as a pH indicator, it is used widely in culture media to identify changes from neutral to acidic pH values

$40.16 - $40.16

Chemical Identifiers

CAS 34487-61-1
Physical Form Shiny Powder
Molecular Formula C19H13O5SNa
Formula Weight 376.4
MDL Number MFCD00066901
Synonym Phenolsulfonphthalein
Chemical Name or Material Phenol Red Sodium Salt
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN10260405 Mp Biomedicals
10260405
5g Each for $40.16
Specifications Description & Specifications

Chemical Identifiers

34487-61-1
C19H13O5SNa
MFCD00066901
Phenol Red Sodium Salt
Shiny Powder
376.4
Phenolsulfonphthalein

Specifications

Shiny Powder
Dark red
Phenolsulfonphthalein
34487-61-1
Phenol Red Sodium Salt
376.4
5g
C19H13O5SNa
MFCD00066901

Safety and Handling

EINECSNumber : 252-057-8