Please login to your online account to display your discounted pricing

Phenol Red Indicator, 0.02% (w/v) Aqueous Solution, Ricca Chemical


Quantity 1L
Packaging Plastic Bottle

Manufacturer: ricca chemical company  RIR57000001A

 View more versions of this product

Catalog No. R57000001A

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Quantity 1L
Chemical Name or Material Phenol Red Indicator
Concentration or Composition (by Analyte or Components) 0.02% (w/v) Aqueous Solution
Packaging Plastic Bottle

Specifications

Chemical Name or Material Phenol Red Indicator